2,4-Dichloro-6-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

15986-1G 15986-25G 15986-5G
76883-266EA 84.88 USD
76883-266 76883-268 76883-270
2,4-Dichloro-6-fluorobenzaldehyde ≥97%
2,4-Dichloro-6-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃Cl₂FO
MW: 193.0000 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12404935
CAS Number: 681435-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR