1,3-Dibromo-2-(phenylmethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

15905-25G 15905-5G
76883-242EA 466.8 USD
76883-242 76883-244
1,3-Dibromo-2-(phenylmethoxy)benzene ≥95%
1,3-Dibromo-2-(phenylmethoxy)benzene
Formula: C₁₃H₁₀Br₂O
Storage Temperature: Ambient
MDL Number: MFCD30295982
CAS Number: 122110-76-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR