2,4,5-Trichlorophenylboronic acid ≥95% research grade

Supplier: AOBChem USA

Warning

15875-1G 15875-25G 15875-500MG 15875-5G
76883-226EA 150.65 USD
76883-226 76883-228 76883-230 76883-232
2,4,5-Trichlorophenylboronic acid ≥95% research grade
2,4,5-Trichlorophenylboronic acid
Formula: C₆H₄BCl₃O₂
MW: 225.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09056706
CAS Number: 220210-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR