1-Fluoro-2,3-dimethoxybenzene ≥95% research grade
Supplier: AOBChem USA
|
Warning
|
15867-25G
15867-5G
76883-220EA
630.18
USD
76883-220
76883-222
1-Fluoro-2,3-dimethoxybenzene ≥95% research grade
1-Fluoro-2,3-dimethoxybenzene
Formula:
C₈H₉FO₂ MW: 156.154 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03411451 CAS Number: 394-64-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...