2,3-Difluoro-6-methoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

15802-25G 15802-5G
76883-212EA 421.18 USD
76883-212 76883-214
2,3-Difluoro-6-methoxybenzaldehyde ≥95%
2,3-Difluoro-6-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆F₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD02093968
CAS Number: 187543-87-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR