(2-Chloro-5-trifluoromethoxy-phenyl)-methanol ≥97%

Supplier: AOBChem USA

Warning

15755-1G 15755-25G 15755-5G
76883-202EA 64.72 USD
76883-202 76883-204 76883-206
(2-Chloro-5-trifluoromethoxy-phenyl)-methanol ≥97%
(2-Chloro-5-trifluoromethoxy-phenyl)-methanol
Formula: C₈H₆ClF₃O₂
MW: 226.58 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18393816
CAS Number: 1261483-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR