1-Bromo-3-propoxybenzene ≥97% research grade
Supplier: AOBChem USA
|
Warning
|
15699-1G
15699-25G
15699-5G
76883-190EA
85.94
USD
76883-190
76883-192
76883-194
1-Bromo-3-propoxybenzene ≥97% research grade
1-Bromo-3-propoxybenzene
Formula:
C₉H₁₁BrO MW: 215.087 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09944153 CAS Number: 149557-17-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...