(2,3-Dichloro-6-fluorophenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,3-Dichloro-6-fluorophenylboronic acid

15632-1G 15632-25G 15632-5G
76883-146EA 96.55 USD
76883-146 76883-148 76883-150
(2,3-Dichloro-6-fluorophenyl)boronic acid ≥97%
(2,3-Dichloro-6-fluorophenyl)boronic acid
Formula: C₆H₄BCl₂FO₂
MW: 208.81 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11111907
CAS Number: 1451393-17-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR