2-(2',4',6'-Trifluorophenoxy)propane ≥97%

Supplier: AOBChem USA

Warning

15471-25G 15471-5G
76883-082EA 346.92 USD
76883-082 76883-084
2-(2',4',6'-Trifluorophenoxy)propane ≥97%
2-(2',4',6'-Trifluorophenoxy)propane
Formula: C₉H₉F₃O
Storage Temperature: Ambient
MDL Number: MFCD19105349
CAS Number: 1310416-65-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR