2,4-Dibromo-6-chlorophenol ≥95%

Supplier: AOBChem USA

Warning

15392-1G 15392-25G 15392-500MG 15392-5G
76883-056EA 168.69 USD
76883-056 76883-058 76883-060 76883-062
2,4-Dibromo-6-chlorophenol ≥95%
2,4-Dibromo-6-chlorophenol

Precaución: for Research Use Only

Formula: C₆H₃Br₂ClO
Storage Temperature: Refrigerator
MDL Number: MFCD00029692
CAS Number: 4526-56-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR