(2,3-Dichloro-5-fluorophenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,3-Dichloro-5-fluorophenylboronic acid

15202-1G 15202-25G 15202-500MG 15202-5G
76882-992EA 260.99 USD
76882-992 76882-994 76882-996 76882-998
(2,3-Dichloro-5-fluorophenyl)boronic acid ≥97%
(2,3-Dichloro-5-fluorophenyl)boronic acid
Formula: C₆H₄BCl₂FO₂
MW: 208.8102 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28786328
CAS Number: 2828433-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR