(2-Chloro-4-fluoro-5-methylphenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

15090-1G 15090-25G 15090-500MG 15090-5G
76882-962EA 203.7 USD
76882-962 76882-964 76882-966 76882-968
(2-Chloro-4-fluoro-5-methylphenyl)boronic acid ≥97% research grade
(2-Chloro-4-fluoro-5-methylphenyl)boronic acid
Formula: C₇H₇BClFO₂
Storage Temperature: Ambient
CAS Number: 1207428-93-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR