(2-Chloro-4-methoxy-5-methylphenyl)boronic acid 95%

Supplier: AOBChem USA

Warning

15088-1G 15088-500MG 15088-5G 15088-25G
76882-948EA 831.75 USD
76882-948 76882-950 76882-952 77653-610
(2-Chloro-4-methoxy-5-methylphenyl)boronic acid 95%
(2-Chloro-4-methoxy-5-methylphenyl)boronic acid
Formula: C₈H₁₀BClO₃
MW: 200.427 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09998844
CAS Number: 502159-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR