2-Piperidino-5-pyrimidineboronic acid pinacol ester ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2-(Piperidin-1-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine, 2-(Piperiden-1-yl)pyrimidine-5-boronic acid, pinacol ester

15021-1G 15021-25G 15021-500MG 15021-5G
76882-934EA 126.25 USD
76882-934 76882-936 76882-938 76882-940
2-Piperidino-5-pyrimidineboronic acid pinacol ester ≥97% research grade
2-Piperidino-5-pyrimidineboronic acid pinacol ester
Formula: C₁₅H₂₄BN₃O₂
MW: 289.1810 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07368250
CAS Number: 1015242-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR