2,3-Difluoro-4-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

14885-1G 14885-25G 14885-5G
76882-914EA 62.6 USD
76882-914 76882-916 76882-918
2,3-Difluoro-4-methoxybenzaldehyde ≥97%
2,3-Difluoro-4-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD04115981
CAS Number: 256417-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR