1-Bromo-4-cyclopentylbenzene ≥97%

Supplier: AOBChem USA

Warning

14788-1G 14788-25G 14788-500MG 14788-5G
76882-894EA 203.7 USD
76882-894 76882-896 76882-898 76882-900
1-Bromo-4-cyclopentylbenzene ≥97%
1-Bromo-4-cyclopentylbenzene
Formula: C₁₁H₁₃Br
Storage Temperature: Refrigerator
MDL Number: MFCD11111741
CAS Number: 59734-91-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR