2,3-Difluoro-4-ethoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

14779-25G 14779-5G
76882-882EA 204.76 USD
76882-882 76882-884
2,3-Difluoro-4-ethoxyphenylboronic acid ≥97%
2,3-Difluoro-4-ethoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₉BF₂O₃
Storage Temperature: Ambient
MDL Number: MFCD09258743
CAS Number: 212386-71-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR