1-Bromo-4-(4-(Methylsulfonyl)Phenoxy)Benzene ≥97%

Supplier: AOBChem USA

Warning

146610-1G 146610-500MG 146610-5G 146610-25G
76882-872EA 555.92 USD
76882-872 76882-874 76882-876 77653-582
1-Bromo-4-(4-(Methylsulfonyl)Phenoxy)Benzene ≥97%
1-Bromo-4-(4-(Methylsulfonyl)Phenoxy)Benzene

Precaución: for Research Use Only

Formula: C₁₃H₁₁BrO₃S
MW: 327.1936 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09832517
CAS Number: 83642-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR