1-Bromo-2-ethoxynaphthalene ≥95%

Supplier: AOBChem USA

Warning

14570-25G 14570-5G
76882-862EA 595.17 USD
76882-862 76882-864
1-Bromo-2-ethoxynaphthalene ≥95%
1-Bromo-2-ethoxynaphthalene
Formula: C₁₂H₁₁BrO
MW: 251.119 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12776103
CAS Number: 50389-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR