2-(2-Bromo-5-methoxyphenyl)-1,3-dioxolane 95%

Supplier: AOBChem USA

Warning

14538-1G 14538-25G 14538-5G 14538-500MG
76882-848EA 112.46 USD
76882-848 76882-850 76882-852 77644-626
2-(2-Bromo-5-methoxyphenyl)-1,3-dioxolane 95%
2-(2-Bromo-5-methoxyphenyl)-1,3-dioxolane
Formula: C₁₀H₁₁BrO₃
MW: 259.097 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06209830
CAS Number: 98015-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR