(E)-4-((hydroxyimino)methyl)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

145285-1G 145285-25G 145285-5G 145285-500MG
76882-834EA 117.76 USD
76882-834 76882-836 76882-838 77648-672
(E)-4-((hydroxyimino)methyl)benzonitrile ≥97%
(E)-4-((hydroxyimino)methyl)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₆N₂O
Storage Temperature: Ambient
MDL Number: MFCD01205660
CAS Number: 52707-54-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR