2-(2-Bromo-4-methylphenyl)-1,3-dioxolane ≥95%

Supplier: AOBChem USA

Warning

14493-1G 14493-25G 14493-5G
76882-814EA 62.6 USD
76882-814 76882-816 76882-818
2-(2-Bromo-4-methylphenyl)-1,3-dioxolane ≥95%
2-(2-Bromo-4-methylphenyl)-1,3-dioxolane
Formula: C₁₀H₁₁BrO₂
MW: 243.097 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06209652
CAS Number: 773095-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR