2-Benzyloxy-5-(trifluoromethyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-(Benzyloxy)-5-(trifluoromethyl)phenylboronic acid

14424-1G 14424-25G 14424-5G
76882-804EA 62.6 USD
76882-804 76882-806 76882-808
2-Benzyloxy-5-(trifluoromethyl)phenylboronic acid ≥97%
2-Benzyloxy-5-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₂BF₃O₃
Storage Temperature: Ambient
MDL Number: MFCD06409944
CAS Number: 612833-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR