1-Benzyloxy-5-bromo-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

14238-25G 14238-5G
76882-786EA 201.58 USD
76882-786 76882-788
1-Benzyloxy-5-bromo-2-methylbenzene ≥97%
1-Benzyloxy-5-bromo-2-methylbenzene
Formula: C₁₄H₁₃BrO
MW: 277.156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11520656
CAS Number: 1114808-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR