1-Bromo-4-((4-methoxybenzyl)oxy)benzene ≥97%

Supplier: AOBChem USA

Warning

141321-1G 141321-25G 141321-5G 141321-500MG
76882-760EA 90.18 USD
76882-760 76882-762 76882-764 77644-610
1-Bromo-4-((4-methoxybenzyl)oxy)benzene ≥97%
1-Bromo-4-((4-methoxybenzyl)oxy)benzene

Precaución: for Research Use Only

Formula: C₁₄H₁₃BrO₂
MW: 293.1558 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11127623
CAS Number: 16201-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR