2-(4-Chlorophenyl)-2-methyl-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

14085-1G 14085-25G 14085-5G
76882-748EA 62.6 USD
76882-748 76882-750 76882-752
2-(4-Chlorophenyl)-2-methyl-1,3-dioxolane ≥97%
2-(4-Chlorophenyl)-2-methyl-1,3-dioxolane
Formula: C₁₀H₁₁ClO₂
Storage Temperature: Refrigerator
MDL Number: MFCD19348735
CAS Number: 36881-02-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR