2-(2-Bromo-4-fluorophenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

14034-1G 14034-25G 14034-500MG 14034-5G
76882-732EA 155.96 USD
76882-732 76882-734 76882-736 76882-738
2-(2-Bromo-4-fluorophenyl)-1,3-dioxolane ≥97%
2-(2-Bromo-4-fluorophenyl)-1,3-dioxolane
Formula: C₉H₈BrFO₂
MW: 247.06 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06209808
CAS Number: 773097-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR