1-Bromo-2-(methoxymethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

13960-1G 13960-25G 13960-5G
76882-710EA 76.39 USD
76882-710 76882-712 76882-714
1-Bromo-2-(methoxymethoxy)benzene ≥97%
1-Bromo-2-(methoxymethoxy)benzene
Formula: C₈H₉BrO₂
MW: 217.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21101767
CAS Number: 68314-54-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR