4-Bromo-3-chloro-2-fluoroiodobenzene ≥97%

Supplier: AOBChem USA

Warning

13901-25G 13901-5G
76882-702EA 254.62 USD
76882-702 76882-704
4-Bromo-3-chloro-2-fluoroiodobenzene ≥97%
4-Bromo-3-chloro-2-fluoroiodobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrClFI
Storage Temperature: Ambient
MDL Number: MFCD09878167
CAS Number: 1000573-03-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR