2-(Oxazol-5-yl)phenol ≥97%

Supplier: AOBChem USA

Warning

13739-1G 13739-25G 13739-500MG 13739-5G
76882-674EA 236.59 USD
76882-674 76882-676 76882-678 76882-680
2-(Oxazol-5-yl)phenol ≥97%
2-(Oxazol-5-yl)phenol
Formula: C₉H₇NO₂
MW: 161.157 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24729988
CAS Number: 391927-03-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR