(2,3-Dichlorophenyl)(phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

136140-1G 136140-25G 136140-500MG 136140-5G
76882-662EA 308.73 USD
76882-662 76882-664 76882-666 76882-668
(2,3-Dichlorophenyl)(phenyl)methanol ≥97%
(2,3-Dichlorophenyl)(phenyl)methanol
Formula: C₁₃H₁₀Cl₂O
MW: 253.1239 g/mol
Storage Temperature: Refrigerator
CAS Number: 286947-77-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR