4-Bromo-1-fluoro-2-naphthaleneboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-Bromo-1-fluoronaphthalen-2-yl)boronic acid

13606-1G 13606-25G 13606-500MG 13606-5G
76882-646EA 124.13 USD
76882-646 76882-648 76882-650 76882-652
4-Bromo-1-fluoro-2-naphthaleneboronic acid ≥97%
4-Bromo-1-fluoro-2-naphthaleneboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₇BBrFO₂
MW: 268.8748 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09027236
CAS Number: 913836-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR