(2-Bromo-6-fluorophenyl)(thiophen-2-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

135279-1G 135279-25G 135279-500MG 135279-5G
76882-630EA 233.4 USD
76882-630 76882-632 76882-634 76882-636
(2-Bromo-6-fluorophenyl)(thiophen-2-yl)methanol ≥95%
(2-Bromo-6-fluorophenyl)(thiophen-2-yl)methanol
Formula: C₁₁H₈BrFOS
MW: 287.148 g/mol
Storage Temperature: Refrigerator
CAS Number: 1592995-43-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR