(2-Fluoro-3-isopropyl-6-methoxyphenyl)boronic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
129721-1G
129721-500MG
129721-5G
129721-25G
76882-576EA
959.06
USD
76882-576
76882-578
76882-580
77653-430
(2-Fluoro-3-isopropyl-6-methoxyphenyl)boronic acid ≥97%
(2-Fluoro-3-isopropyl-6-methoxyphenyl)boronic acid
Precaución: for Research Use Only
Formula:
C₁₀H₁₄BFO₃ MW: 212.026 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34828221 CAS Number: 3054532-18-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...