(3-Fluorophenyl)(furan-2-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

128303-1G 128303-25G 128303-500MG 128303-5G
76882-520EA 332.07 USD
76882-520 76882-522 76882-524 76882-526
(3-Fluorophenyl)(furan-2-yl)methanol ≥97%
(3-Fluorophenyl)(furan-2-yl)methanol
Formula: C₁₁H₉FO₂
MW: 192.186 g/mol
Storage Temperature: Refrigerator
CAS Number: 944684-11-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR