2,4,6-Trifluoroiodobenzene ≥97%

Supplier: AOBChem USA

Warning

12611-25G 12611-5G
76882-460EA 289.63 USD
76882-460 76882-462
2,4,6-Trifluoroiodobenzene ≥97%
2,4,6-Trifluoroiodobenzene
Formula: C₆H₂F₃I
Storage Temperature: Ambient
MDL Number: MFCD03095328
CAS Number: 41860-63-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR