2,4,6-Trimethoxyphenylboronic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
12552-1G
12552-25G
12552-5G
76882-450EA
84.88
USD
76882-450
76882-452
76882-454
2,4,6-Trimethoxyphenylboronic acid ≥97%
2,4,6-Trimethoxyphenylboronic acid
Precaución: for Research Use Only
Formula:
C₉H₁₃BO₅ Storage Temperature: Refrigerator |
MDL Number:
MFCD01114642 CAS Number: 135159-25-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...