1-Bromo-4-Methyl-2-(Methylsulfonyl)Benzene ≥97%

Supplier: AOBChem USA

Warning

123771-1G 123771-25G 123771-500MG 123771-5G
76882-416EA 201.58 USD
76882-416 76882-418 76882-420 76882-422
1-Bromo-4-Methyl-2-(Methylsulfonyl)Benzene ≥97%
1-Bromo-4-Methyl-2-(Methylsulfonyl)Benzene
Formula: C₈H₉BrO₂S
MW: 249.125 g/mol
Storage Temperature: Ambient
CAS Number: 254897-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR