(2-(cyclopentyloxy)pyrimidin-5-yl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

121741-1G 121741-25G 121741-500MG 121741-5G
76882-380EA 145.35 USD
76882-380 76882-382 76882-384 76882-386
(2-(cyclopentyloxy)pyrimidin-5-yl)boronic acid ≥97%
(2-(cyclopentyloxy)pyrimidin-5-yl)boronic acid
Formula: C₉H₁₃BN₂O₃
MW: 208.022 g/mol
Storage Temperature: Refrigerator
CAS Number: 2225155-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR