1,2-Dibromo-3,5-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

121709-1G 121709-25G 121709-500MG 121709-5G
76882-364EA 233.4 USD
76882-364 76882-366 76882-368 76882-370
1,2-Dibromo-3,5-dimethylbenzene ≥97%
1,2-Dibromo-3,5-dimethylbenzene
Formula: C₈H₈Br₂
MW: 263.957 g/mol
Storage Temperature: Ambient
CAS Number: 116668-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR