2,3-Difluoro-6-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

12145-25G 12145-5G
76882-356EA 346.92 USD
76882-356 76882-358
2,3-Difluoro-6-methoxyphenylboronic acid ≥97%
2,3-Difluoro-6-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BF₂O₃
MW: 187.94 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03095116
CAS Number: 957061-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR