1-(3-Bromo-2-fluorophenyl)propan-1-ol ≥97%

Supplier: AOBChem USA

Warning

119971-1G 119971-25G 119971-500MG 119971-5G
76882-326EA 233.4 USD
76882-326 76882-328 76882-330 76882-332
1-(3-Bromo-2-fluorophenyl)propan-1-ol ≥97%
1-(3-Bromo-2-fluorophenyl)propan-1-ol
Formula: C₉H₁₀BrFO
Storage Temperature: Refrigerator
CAS Number: 1699539-87-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR