(2-((Dimethylamino)methyl)phenyl)boronic acid ≥95% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2-(N,N-Dimethylaminomethyl)phenylboronic acid, 2-(Dimethylaminomethyl)phenylboronic acid, 2-(Dimethylaminomethyl)benzeneboronic acid

11854-25G 11854-5G
76882-310EA 283.27 USD
76882-310 76882-312
(2-((Dimethylamino)methyl)phenyl)boronic acid ≥95% research grade
(2-((Dimethylamino)methyl)phenyl)boronic acid
Formula: C₉H₁₄BNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD01318999
CAS Number: 85107-53-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR