2,4-Dibromo-5-fluoroaniline ≥97%

Supplier: AOBChem USA

Warning

11816-25G 11816-5G
76882-288EA 488.02 USD
76882-288 76882-290
2,4-Dibromo-5-fluoroaniline ≥97%
2,4-Dibromo-5-fluoroaniline

Precaución: for Research Use Only

Formula: C₆H₄Br₂FN
Storage Temperature: Ambient
MDL Number: MFCD09878124
CAS Number: 1000578-04-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR