2-(4-Fluorophenyl)benzo[d]thiazole ≥97% research grade

Supplier: AOBChem USA

Warning

117261-1G 117261-25G 117261-500MG 117261-5G
76882-274EA 144.29 USD
76882-274 76882-276 76882-278 76882-280
2-(4-Fluorophenyl)benzo[d]thiazole ≥97% research grade
2-(4-Fluorophenyl)benzo[d]thiazole
Formula: C₁₃H₈FNS
Storage Temperature: Ambient
CAS Number: 1629-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR