2-Piperidino-5-pyridineboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-(Piperidin-1-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-(Piperidin-1-yl)pyridine-5-boronic acid pinacol ester

11649-25G 11649-5G
76882-232EA 595.17 USD
76882-232 76882-234
2-Piperidino-5-pyridineboronic acid pinacol ester ≥97%
2-Piperidino-5-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₆H₂₅BN₂O₂
MW: 288.1929 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06798274
CAS Number: 852228-08-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR