2-(4-Fluoro-2-(trifluoromethyl)phenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

116289-25G 116289-5G 116289-1G 116289-500MG
76882-228EA 112.46 USD
76882-228 77631-060 77636-372 77652-290
2-(4-Fluoro-2-(trifluoromethyl)phenyl)-1,3-dioxolane ≥97%
2-(4-Fluoro-2-(trifluoromethyl)phenyl)-1,3-dioxolane

Precaución: for Research Use Only

Formula: C₁₀H₈F₄O₂
MW: 236.163 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06209163
CAS Number: 773102-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR