(2-((4-Methylbenzyl)oxy)phenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

114250-1G 114250-25G 114250-500MG 114250-5G
76882-182EA 357.53 USD
76882-182 76882-184 76882-186 76882-188
(2-((4-Methylbenzyl)oxy)phenyl)boronic acid ≥97% research grade
(2-((4-Methylbenzyl)oxy)phenyl)boronic acid
Formula: C₁₄H₁₅BO₃
Storage Temperature: Ambient
CAS Number: 1311182-72-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR