1-Bromo-4-isopropoxybenzene ≥97%
Supplier: AOBChem USA
|
Warning
|
11183-25G
11183-5G
76882-150EA
204.76
USD
76882-150
76882-152
1-Bromo-4-isopropoxybenzene ≥97%
1-Bromo-4-isopropoxybenzene
Formula:
C₉H₁₁BrO Storage Temperature: Ambient |
MDL Number:
MFCD00089972 CAS Number: 6967-88-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...