1-Benzyloxy-4-bromo-2-chlorobenzene ≥97% research grade

Supplier: AOBChem USA

Warning

11128-25G 11128-5G
76882-128EA 310.85 USD
76882-128 76882-130
1-Benzyloxy-4-bromo-2-chlorobenzene ≥97% research grade
1-Benzyloxy-4-bromo-2-chlorobenzene
Formula: C₁₃H₁₀BrClO
MW: 297.5800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09265138
CAS Number: 56872-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR